Merck
  • Environmental implications and applications of carbon nanomaterials in water treatment.
Environmental implications and applications of carbon nanomaterials in water treatment.
Water science and technology : a journal of the International Association on Water Pollution Research (2013-06-12)
S R Chae, E M Hotze, A R Badireddy, S Lin, J O Kim, M R Wiesner
ABSTRACT
Carbon nanomaterials have been proposed as a basis for developing new technologies for photocatalytic oxidation and disinfection, improved membrane processes, adsorbents, and biofilm-resistant surfaces. This study details recent progress towards the development of these proposed applications. We explored the use of carbon nanomaterials such as fullerene C60, single-wall carbon nanotubes (SWCNTs), and multi-wall carbon nanotubes (MWCNTs) for a range of new technologies including, degradation of a probe organic compound by in situ generation of reactive oxygen species (ROS), new strategies for microbial disinfection, and the inhibition of biofilm development on membrane surfaces. The results show that the degradation of 2-chlorophenol by ROS produced microbial inactivation, and the mobility of the nanoparticle aggregates of the carbon nanomaterials all increased as suspensions were fractionated to enrich with smaller aggregates with sonication followed by successive membrane filtration.
Sigma-Aldrich
Silica, nanopowder, 99.8% trace metals basis
Sigma-Aldrich
Silica
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silicon dioxide, alumina doped, nanoparticles, dispersion, <50 nm particle size, 20 wt. % in H2O, ≥99.9% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Supelco
2-Chlorophenol, PESTANAL®, analytical standard
Sigma-Aldrich
2-Chlorophenol, ≥99%
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
2-Chlorophenol, 98%
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O