Merck
  • Photoinduced ligand exchange and DNA binding of cis-[Ru(phpy)(phen)(CH3CN)2]+ with long wavelength visible light.
Photoinduced ligand exchange and DNA binding of cis-[Ru(phpy)(phen)(CH3CN)2]+ with long wavelength visible light.
Journal of inorganic biochemistry (2013-01-29)
R Bryan Sears, Lauren E Joyce, Maya Ojaimi, Judith C Gallucci, Randolph P Thummel, Claudia Turro
ABSTRACT
The complex cis-[Ru(phpy)(phen)(CH3CN)2](+) (phpy=2-phenylpyridine, phen=1,10-phenanthroline) was investigated as a potential photodynamic therapy (PDT) agent. This complex presents desirable photochemical characteristics including a low energy absorption tail extending into the PDT window (600-850nm) and photoinduced exchange of the CH3CN ligands, generating a species analogous to the chemotherapy drug cisplatin. Furthermore, photochemical reactivity can be controlled through selective irradiation into the Ru-phen singlet metal-to-ligand charge transfer ((1)MLCT) band (λirr=500 nm) of [Ru(phpy)(phen)(CH3CN)2](+) in the presence of excess t-butylammonium chloride (TBACl) resulting in efficient photoinduced production of [Ru(phpy)(phen)(CH3CN)Cl] (Φ=0.25). This lower energy irradiation resulted in greater quantum yield of photosubstitution when compared to direct irradiation into the Ru-phpy (1)MLCT peak (λirr=450 nm; Φ=0.08) in CH2Cl2. It was found that the lower quantum yield observed for irradiation into the Ru→phpy(-)(1)MLCT band results from significant orbital mixing of the phpy(-) ligand with the t2g-type filled set in the metal, giving this state significant ligand-centered character. Lastly, this complex produced a decrease in the mobility of linearized ds-DNA when irradiated with λirr≥420nm, indicative of covalent binding by the transition metal complex similar to that observed for cisplatin. No change in mobility was found for the same samples kept in the dark indicating, unlike cisplatin, DNA binding of cis-[Ru(phpy)(phen)(CH3CN)2](+) only occurs with the activation of light. These observations support the use of cis-[Ru(phpy)(phen)(CH3CN)2](+) as a potential PDT agent by the photoinduced generation of a cisplatin analog.
Sigma-Aldrich
Tetrabutylammonium cyanide, technical, ≥80%
Supelco
Tetrabutylammonium perchlorate, for electrochemical analysis, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium perchlorate, ≥95.0% (T)
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 53.5-56.5% in H2O
Supelco
Tetrabutylammonium hydroxide solution, ~40% in water, suitable for ion chromatography
Sigma-Aldrich
Tetrabutylammonium cyanide, 95%
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic solution, 1.0 M in H2O
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, 97%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
Sigma-Aldrich
Tetrabutylammonium bromide solution, 50 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium nitrate, 97%
Sigma-Aldrich
Tetrabutylammonium chloride, ≥97.0% (NT)
Supelco
Tetrabutylammonium bisulfate solution, suitable for ion pair chromatography, LiChropur, concentrate, ampule
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic, puriss., 99% (T)
Supelco
Tetrabutylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
Supelco
Tetrabutylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Tetrabutylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
Tetrabutylammonium iodide, reagent grade, 98%
Sigma-Aldrich
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 75 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium azide
Sigma-Aldrich
Tetrabutylammonium iodide, ≥99.0% (AT)
Supelco
Tetrabutylammonium iodide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 1.0 M in THF