Skip to Content
Merck
All Photos(1)

Key Documents

361399

Sigma-Aldrich

Tetrabutylammonium fluoride solution

75 wt. % in H2O

Synonym(s):

TBAF solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]4NF
CAS Number:
Molecular Weight:
261.46
Beilstein:
3762762
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
form:
viscous liquid

form

viscous liquid

Quality Level

concentration

75 wt. % in H2O

density

0.953 g/mL at 25 °C

functional group

amine

SMILES string

[F-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.FH/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H/q+1;/p-1

InChI key

FPGGTKZVZWFYPV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Triple monoamine reuptake inhibitors as a new generation of antidepressants
  • Alcohols via hydrolysis of alkyl silyl ethers neutral pH in mixed organic-aqueous buffered solutions
  • Oligoribonucleotides with phosphonate-modified linkages
  • Aryl alkyl alcohols via Nozaki-Hiyama allylation catalyzed by chiral bipyridyldiol ligands and chromium trichloride
  • Conjugated dienoic acid esters using Suzuki coupling reactions
  • Macrocyclic o-aminobenzamide Hsp90 inhibitorwith antitumor activity
  • Phospshoinositide 3-kinase (PI3K)/mammalian target of rapamycin (mTOR) dual inhibitors
  • Anti-diabetic polyacetylenic glucosides

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oliver Bixner et al.
The Journal of chemical physics, 136(20), 204503-204503 (2012-06-07)
The interaction of exciton and charge transfer (CT) states plays a central role in photo-induced CT processes in chemistry, biology, and physics. In this work, we use a combination of two-dimensional electronic spectroscopy (2D-ES), pump-probe measurements, and quantum chemistry to
Mei Shen et al.
Journal of the American Chemical Society, 134(24), 9856-9859 (2012-06-05)
Here we report on the unprecedentedly high resolution imaging of ion transport through single nanopores by scanning electrochemical microscopy (SECM). The quantitative SECM image of single nanopores allows for the determination of their structural properties, including their density, shape, and
Chao Zhong et al.
Carbohydrate polymers, 94(1), 38-45 (2013-04-03)
In this article, a novel and high efficient solvent, tetra-n-Butylammonium Hydroxide (TBAH), was used for dissolution and isolation of straw cellulose from wheat straw. The composition analysis with gas chromatography (GC) and the spectroscopic characterization analysis conducted by X-Ray diffraction
Morteza Moradi et al.
The Analyst, 137(15), 3549-3557 (2012-06-27)
Hollow fiber-based liquid phase microextraction (HF-LPME) using conventional solvents is limited by their relative instability and high volatility. The use of supramolecular solvents as a liquid membrane phase could overcome these inconveniences due to their negligible vapour pressure and high
Noelia Luque et al.
Journal of chromatography. A, 1248, 74-83 (2012-06-26)
The suitability of a mixed-mode sorbent made up of admicelles of sodium dodecyl sulphate (SDS) and tetrabutylammonium (TBA) to extract and preserve pesticides from river and underground water was assessed. Pesticides belonging to different structural groups (i.e. triazines, carbamates, phenylureas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service