Skip to Content
Merck
All Photos(1)

Key Documents

79989

Sigma-Aldrich

Tetrabutylammonium cyanide

technical, ≥80%

Synonym(s):

N,N,N-tributyl-1-butanaminium cyanide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$1,530.00

$1,530.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$1,530.00

About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(CN)
CAS Number:
Molecular Weight:
268.48
Beilstein:
3598808
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$1,530.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical

Quality Level

Assay

≥80%

form

crystals

mp

89-92 °C (lit.)

SMILES string

[C-]#N.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.CN/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-2/h5-16H2,1-4H3;/q+1;-1

InChI key

KRRBFUJMQBDDPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tetrabutylammonium cyanide can be used as a catalyst:
  • For the deprotection of aliphatic thioacetate to synthesize free thiols in the presence of a protic solvent[1].
  • In the O-TMS cyanosilylation of carbonyl compounds to synthesize cyanohydrin trimethylsilyl ethers in the presence of trimethylsilyl cyanide (TMSCN)[2].
  • For the ring expansion of β-lactams to synthesize γ-lactams through a bond cleavage of the β-lactam in the presence of acetonitrile[3].

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benito Alcaide et al.
Organic letters, 7(18), 3981-3984 (2005-08-27)
Tetrabutylammonium cyanide (20 mol %) catalyzes ring expansion of 4-(arylimino)methylazetidin-2-ones 2 to 5-aryliminopyrrolidin-2-ones 3 through a novel N1-C4 bond cleavage of the beta-lactam nucleus. New, efficient one-pot protocols to enantiopure succinimide derivatives 3 and 4 from beta-lactam aldehydes 1 have
Tetrabutylammonium cyanide catalyzes the addition of TMSCN to aldehydes and ketones
Cordoba R, et al.
ARKIVOC (Gainesville, FL, United States), 4, 94-99 (2004)
Aliphatic thioacetate deprotection using catalytic tetrabutylammonium cyanide
Holmes B, et al.
Tetrahedron, 61, 12339-12342 (2005)
Fabio Galeotti et al.
Journal of chromatography. A, 1284, 141-147 (2013-03-05)
In this study, by using tetrabutylammonium bisulfate as ion-pairing reagent, we were able to separate all the main heparin/heparan sulfate disaccharides generated by the action of heparinases along with the main Hep tetrasaccharide possessing a 3-O-sulfate group on the sulfoglucosamine
Erbo Shi et al.
Organic letters, 14(13), 3384-3387 (2012-06-27)
A metal-free C-H oxidation for the construction of allylic esters has been developed. The use of a commercially available and inexpensive catalyst and oxidant, and readily available starting materials, coupled with the operational simplicity of the reaction, renders the methodology

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service