Skip to Content
Merck
All Photos(2)

Key Documents

178780

Sigma-Aldrich

Tetrabutylammonium hydroxide solution

greener alternative

40 wt. % in H2O

Synonym(s):

N,N,N-Tributyl-1-butanaminium hydroxide, TBAH 40, Tetra-n-butylammonium hydroxide

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$191.00
250 ML
$604.00
1 L
$1,830.00
2.5 L
$3,270.00

$191.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
50 ML
$191.00
250 ML
$604.00
1 L
$1,830.00
2.5 L
$3,270.00

About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(OH)
CAS Number:
Molecular Weight:
259.47
Beilstein:
3571228
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
concentration:
40 wt. % in H2O

$191.00


In StockDetails


Request a Bulk Order

form

liquid

Quality Level

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

40 wt. % in H2O

refractive index

n20/D 1.405

density

0.99 g/mL at 25 °C

functional group

amine

greener alternative category

SMILES string

[OH-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.H2O/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H2/q+1;/p-1

InChI key

VDZOOKBUILJEDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetrabutylammonium hydroxide is a quaternary ammonium salt mainly used as a strong base and phase-transfer catalyst.[1] It can be synthesized from tetrabutylammonium iodide.[2]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is highly effcient and enhance catalytic efficiency. Click here for more information.

Application

Tetrabutylammonium hydroxide solution (TBAOH) may be used in the following studies:
  • For the dissolution of 10wt% cellulose under room temperature conditions.[3]
  • As a catalyst during the synthesis of 1,2,4-oxadiazoles.[4]
  • Preparation of TBAOH salts of fatty acids.[5]

Caution

May crystallize below 30°C
Gentle warming and swirling should redissolve the solid

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrabutylammonium Hydroxide
Bos ME.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2001)
Tetrabutylammonium Hydroxide as Titrant for Acids in Nonaqueous Solutions.
Cundiff RH and Markunas PC.
Analytical Chemistry, 28(5), 792-797 (1956)
B B Y Lau et al.
Bioresource technology, 197, 252-259 (2015-09-08)
Aqueous solutions of tetrabutylphosphonium hydroxide have been evaluated as pretreatment media for rice husks, prior to sulphuric acid hydrolysis or cellulase enzymatic hydrolysis. Varying the water:tetrabutylphosphonium hydroxide ratio varied the rate of delignification, as well as silica, lignin and cellulose
Foaming and emulsifying properties of fatty acids neutralized by tetrabutylammonium hydroxide.
Fameau AL, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 403, 87-95 (2012)
Construction of 3, 5-substituted 1, 2, 4-oxadiazole rings triggered by tetrabutylammonium hydroxide: a highly efficient and fluoride-free ring closure reaction of O-acylamidoximes.
Otaka H, et al.
Tetrahedron Letters, 55(5), 979-981 (2014)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service