Merck
  • Effects of sodium carbonate pretreatment on the chemical compositions and enzymatic saccharification of rice straw.
Effects of sodium carbonate pretreatment on the chemical compositions and enzymatic saccharification of rice straw.
Bioresource technology (2012-09-20)
Linfeng Yang, Jie Cao, Yongcan Jin, Hou-min Chang, Hasan Jameel, Richard Phillips, Zhongzheng Li
ABSTRACT
The effects of sodium carbonate (Na(2)CO(3)) pretreatment on the chemical compositions and enzymatic saccharification of rice straw were investigated. The enzymatic digestibility of rice straw is enhanced after pretreatment since pretreated solids show significant delignification with high sugar availability. During pretreatment, an increasing temperature and Na(2)CO(3) charge leads to enhanced delignification, whereas an increased degradation of polysaccharides as well, of which xylan acts more susceptible than glucan. The sugar recovery of enzymatic hydrolysis goes up rapidly with the total titratable alkali (TTA) increasing from 0% to 8%, and then it reaches a plateau. The highest sugar recovery of rice straw after pretreatment, 71.7%, 73.2%, and 76.1% for total sugar, glucan, and xylan, respectively, is obtained at 140°C, TTA 8% and cellulase loading of 20 FPU/g-cellulose. In this condition, the corresponding delignification ratio of pretreated solid is 41.8%, while 95% of glucan and 76% of xylan are conserved.
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate decahydrate, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium carbonate monohydrate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate monohydrate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, 99.5-100.5% (ex dried subst.)
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate decahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate monohydrate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Supelco
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7