Skip to Content
Merck
All Photos(4)

Key Documents

230952

Sigma-Aldrich

Sodium carbonate monohydrate

ACS reagent, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3 · H2O
CAS Number:
Molecular Weight:
124.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
powder or crystals
Pricing and availability is not currently available.

grade

ACS reagent

Assay

≥99.5%

form

powder or crystals

impurities

≤0.001% N compounds
≤0.004% S compounds
≤0.005% silica
≤0.01% NH4OH ppt.
≤0.01% insolubles

loss

13.0-15.0% loss on drying

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤5 ppm
sulfur compounds (as SO42-): ≤0.004%

cation traces

Ca: ≤0.03%
Fe: ≤5 ppm
K: ≤0.005%
Mg: ≤0.005%
heavy metals (as Pb): ≤5 ppm

SMILES string

O.[Na+].[Na+].[O-]C([O-])=O

InChI

1S/CH2O3.2Na.H2O/c2-1(3)4;;;/h(H2,2,3,4);;;1H2/q;2*+1;/p-2

InChI key

MQRJBSHKWOFOGF-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Buffer component, may be used for the removal of peripheral membrane proteins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dehydration of sodium carbonate monohydrate.
Ball MC, et al.
Journal of the Chemical Society, Faraday Transactions 1: Physical Chemistry in Condensed Phases, 81(8), 1761-1766 (1985)
Eagleson M.
Concise Encyclopedia Chemistry, 993-994 (1994)
Dehydration of sodium carbonate monohydrate with indirect microwave heating.
Seyrankaya A and Ozalp B.
Thermochimica Acta, 448(1), 31-36 (2006)
Azadeh Nilghaz et al.
Lab on a chip, 12(1), 209-218 (2011-11-18)
This paper describes the fabrication of microfluidic cloth-based analytical devices (μCADs) using a simple wax patterning method on cotton cloth for performing colorimetric bioassays. Commercial cotton cloth fabric is proposed as a new inexpensive, lightweight, and flexible platform for fabricating
Yali Zhang et al.
Journal of hazardous materials, 213-214, 167-174 (2012-02-16)
Cyanide tailing is a kind of solid waste produced in the process of gold extraction from gold ore. In this paper, recovery of iron from cyanide tailings was studied with reduction roasting-water leaching process followed by magnetic separation. After analysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service