Merck
  • Convenient and efficient approach to the permanent or reversible conjugation of RNA and DNA sequences with functional groups.
Convenient and efficient approach to the permanent or reversible conjugation of RNA and DNA sequences with functional groups.
Current protocols in nucleic acid chemistry (2012-09-08)
Jacek Cieślak, Cristina Ausín, Andrzej Grajkowski, Serge L Beaucage
ABSTRACT
The conversion of 3',5'-disilylated 2'-O-(methylthiomethyl)ribonucleosides to 2'-O-(phthalimidooxymethyl)ribonucleosides is achieved in yields of 66% to 94%. Desilylation and dephtalimidation of these ribonucleosides by treatment with NH(4)F in MeOH produce 2'-O-aminooxymethylated ribonucleosides, which are efficient in producing stable and yet reversible 2'-conjugates upon reaction with 1-pyrenecarboxaldehyde. Exposure of 2'-pyrenylated ribonucleosides to 0.5 M tetra-n-butylammonium fluoride (TBAF) in THF or DMSO results in the cleavage of their iminoether functions to give the native ribonucleosides along with an innocuous nitrile side product. Conversely, the reaction of 2'-O-(aminooxymethyl)uridine with 5-cholesten-3-one leads to a permanent uridine 2'-conjugate, which is left unreacted when treated with TBAF. The versatility and uniqueness of 2'-O-(aminooxymethyl)ribonucleosides is demonstrated by the single or double incorporation of a reversible pyrenylated uridine 2'-conjugate into an RNA sequence. Furthermore, the conjugation of 2'-O-(aminooxymethyl)ribonucleosides with various aldehydes, including those generated from their acetals, is also presented. The preparation of 5'-O-(aminooxymethyl)thymidine is also achieved, albeit in modest yields, from the conversion of 5'-O-methylthiomethyl-3'-O-(levulinyl)thymidine to 5'-O-phthalimidooxymethyl-3'-O-(levuliny)lthymidine followed by hydrazinolysis of both 5'-phthalimido and 3'-levulinyl groups. Pyrenylation of the 5'-O-(aminooxymethyl)deoxyribonucleoside also provides a reversible 5'-conjugate that is sensitive to TBAF, thereby further demonstrating the usefulness of 5'-O-(aminooxymethyl)deoxyribonucleosides for permanent or reversible modification of DNA sequences. Curr. Protoc. Nucleic Acid Chem. 50:4.52.1-4.52.36. © 2012 by John Wiley & Sons, Inc.
Product Description
Supelco
Tetrabutylammonium bisulfate solution, suitable for ion pair chromatography, LiChropur, concentrate, ampule
Supelco
Tetrabutylammonium iodide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium iodide, ≥99.0% (AT)
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 1.0 M in THF
Sigma-Aldrich
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium bromide solution, 50 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium iodide, reagent grade, 98%
Sigma-Aldrich
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 75 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium nitrate, 97%
Sigma-Aldrich
Tetrabutylammonium chloride, ≥97.0% (NT)
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic, puriss., 99% (T)
Supelco
Tetrabutylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Tetrabutylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
Supelco
Tetrabutylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium azide
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
Supelco
Tetrabutylammonium hydroxide solution, ~40% in water, suitable for ion chromatography
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 53.5-56.5% in H2O
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic solution, 1.0 M in H2O
Sigma-Aldrich
Tetrabutylammonium cyanide, 95%
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, 97%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
Sigma-Aldrich
Tetrabutylammonium perchlorate, ≥95.0% (T)
Supelco
Tetrabutylammonium perchlorate, for electrochemical analysis, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium cyanide, technical, ≥80%