Merck
  • Adsorption of phenanthrene by quaternary ammonium surfactant modified peat and the mechanism involved.
Adsorption of phenanthrene by quaternary ammonium surfactant modified peat and the mechanism involved.
Water science and technology : a journal of the International Association on Water Pollution Research (2012-07-07)
Y B Zhou, L Chen, X Q Wang, Y X Xu, J Lu
ABSTRACT
Removal of phenanthrene (PHE) from aqueous solution by adsorption onto quaternary ammonium surfactant modified peat was studied. The results show that surfactant modification enhanced the PHE adsorption capacity of peat. Low temperature and neutral pH favored PHE adsorption. Peat modified with long carbon chain surfactant performed better than peat modified with short carbon chain surfactant. The magnitude of PHE adsorption capacity followed the order of MP-HPB>MP-HTAB>MP-TBAB>RP, ranged from 924 to 1,228 μg g(-1). A negative trend between adsorption capacity (y) and (O+N)/C ratio of biosorbent (x) was observed (y = -1,369.6x + 2,176), which confirmed the negative effect of polarity on polycyclic aromatic hydrocarbon (PAH) removal. The study provides a guide to modify raw materials to enhance adsorption of hydrophobic organics.
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic solution, 1.0 M in H2O
Sigma-Aldrich
Tetrabutylammonium cyanide, 95%
Sigma-Aldrich
Tetrabutylammonium perchlorate, ≥95.0% (T)
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
Supelco
Tetrabutylammonium perchlorate, for electrochemical analysis, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 53.5-56.5% in H2O
Supelco
Tetrabutylammonium hydroxide solution, ~40% in water, suitable for ion chromatography
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, 97%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
Sigma-Aldrich
Tetrabutylammonium cyanide, technical, ≥80%
Supelco
Tetrabutylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic, puriss., 99% (T)
Supelco
Tetrabutylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Tetrabutylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 1.0 M in THF
Sigma-Aldrich
Tetrabutylammonium iodide, reagent grade, 98%
Sigma-Aldrich
Tetrabutylammonium bromide solution, 50 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium chloride, ≥97.0% (NT)
Supelco
Tetrabutylammonium bisulfate solution, suitable for ion pair chromatography, LiChropur, concentrate, ampule
Sigma-Aldrich
Tetrabutylammonium iodide, ≥99.0% (AT)
Supelco
Tetrabutylammonium iodide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O
Sigma-Aldrich
Tetrabutylammonium nitrate, 97%
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 75 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
Tetrabutylammonium azide
Sigma-Aldrich
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)