Merck
  • Notch receptor-ligand binding facilitates extracellular vesicle-mediated neuron-to-neuron communication.
Notch receptor-ligand binding facilitates extracellular vesicle-mediated neuron-to-neuron communication.
Cell reports (2024-01-19)
Yi-Zhi Wang, Charlotte C M Castillon, Kamil K Gebis, Elizabeth T Bartom, Alessandra d'Azzo, Anis Contractor, Jeffrey N Savas
ABSTRACT
Extracellular vesicles (EVs) facilitate intercellular communication by transferring cargo between cells in a variety of tissues. However, how EVs achieve cell-type-specific intercellular communication is still largely unknown. We found that Notch1 and Notch2 proteins are expressed on the surface of neuronal EVs that have been generated in response to neuronal excitatory synaptic activity. Notch ligands bind these EVs on the neuronal plasma membrane, trigger their internalization, activate the Notch signaling pathway, and drive the expression of Notch target genes. The generation of these neuronal EVs requires the endosomal sorting complex required for transport-associated protein Alix. Adult Alix conditional knockout mice have reduced hippocampal Notch signaling activation and glutamatergic synaptic protein expression. Thus, EVs facilitate neuron-to-neuron communication via the Notch receptor-ligand system in the brain.
Sigma-Aldrich
(+)-Bicuculline, ≥97.0% (TLC)
Sigma-Aldrich
5-Fluoro-2′-deoxyuridine, thymidylate synthase inhibitor
Sigma-Aldrich
H-89 dihydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-LBPA Antibody, clone 6C4, clone 6C4, from mouse
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 1 Antibody, serum, Chemicon®
Sigma-Aldrich
Hydroxy-Dynasore, ≥98% (HPLC)
Sigma-Aldrich
Anti-MAP2 Antibody, Chemicon®, from chicken
Sigma-Aldrich
DL-2-Amino-5-phosphonopentanoic acid, solid
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Picrotoxin, powder
Sigma-Aldrich
KN-93, ≥98% (HPLC)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis