Skip to Content
Merck
All Photos(1)

Key Documents

SML0340

Sigma-Aldrich

Hydroxy-Dynasore

≥98% (HPLC)

Synonym(s):

3-Hydroxy-2-naphthalenecarboxylic acid 2-[(2,4,5-trihydroxyphenyl)methylene]hydrazide, 3-Hydroxy-N′-[(2,4,5-trihydroxyphenyl)methylidene]naphthalene-2-carbohydrazide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$160.00
25 MG
$634.00

$160.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$160.00
25 MG
$634.00

About This Item

Empirical Formula (Hill Notation):
C18H14N2O5
CAS Number:
Molecular Weight:
338.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$160.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

faintly yellow to dark yellow

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

Oc1cc(O)c(\C=N\NC(=O)c2cc3ccccc3cc2O)cc1O

InChI

1S/C18H14N2O5/c21-14-8-17(24)16(23)7-12(14)9-19-20-18(25)13-5-10-3-1-2-4-11(10)6-15(13)22/h1-9,21-24H,(H,20,25)/b19-9+

InChI key

UAXHPUSKEWEOAP-DJKKODMXSA-N

Biochem/physiol Actions

Hydroxy-Dynasore is a cell permeable and potent dynamin inhibitor that prevents uptake of recombinant botulinum neurotoxin A heavy chain binding domain (BoNT/A-Hc). Apparently, Hydroxy-Dynasore prevents dynamin-mediated fission of endocytic vesicles from the plasma membrane.
Hydroxy-Dynasore is a cell permeable and potent dynamin inhibitor.

Other Notes

Air sensitive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thao Nguyen et al.
Oncogene, 38(35), 6283-6300 (2019-07-18)
N-cadherin adhesion has been reported to enhance cancer and neuronal cell migration either by mediating actomyosin-based force transduction or initiating fibroblast growth factor receptor (FGFR)-dependent biochemical signalling. Here we show that FGFR1 reduces N-cadherin-mediated cell migration. Both proteins are co-stabilised
Hui Yi Chew et al.
Cell, 180(5), 895-914 (2020-03-07)
A safe and controlled manipulation of endocytosis in vivo may have disruptive therapeutic potential. Here, we demonstrate that the anti-emetic/anti-psychotic prochlorperazine can be repurposed to reversibly inhibit the in vivo endocytosis of membrane proteins targeted by therapeutic monoclonal antibodies, as directly demonstrated
Keisuke Shirakura et al.
EMBO molecular medicine, 15(4), e16128-e16128 (2023-02-07)
Vascular endothelial protein tyrosine phosphatase (VE-PTP) influences endothelial barrier function by regulating the activation of tyrosine kinase receptor Tie2. We determined whether this action is linked to the development of atherosclerosis by examining the influence of arterial shear stress on
Alekhya Mazumdar et al.
International journal of molecular sciences, 21(15) (2020-08-06)
Tumor-secreted extracellular vesicles (EVs) have been identified as mediators of cancer-host intercellular communication and shown to support pre-metastatic niche formation by modulating stromal cells at future metastatic sites. While osteosarcoma, the most common primary malignant bone tumor in children and
Yi-Zhi Wang et al.
Cell reports, 43(2), 113680-113680 (2024-01-19)
Extracellular vesicles (EVs) facilitate intercellular communication by transferring cargo between cells in a variety of tissues. However, how EVs achieve cell-type-specific intercellular communication is still largely unknown. We found that Notch1 and Notch2 proteins are expressed on the surface of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service