Merck
  • Carbonic anhydrase activators. Activation of the membrane-associated isoform XV with amino acids and amines.
Carbonic anhydrase activators. Activation of the membrane-associated isoform XV with amino acids and amines.
Bioorganic & medicinal chemistry letters (2009-05-26)
Alessio Innocenti, Mika Hilvo, Seppo Parkkila, Andrea Scozzafava, Claudiu T Supuran
ABSTRACT
An activation study of the membrane-associated carbonic anhydrase (CA, EC 4.2.1.1) isoform XV with a series of natural and non-natural amino acids and aromatic/heterocyclic amines is reported. Murine CA XV was strongly activated by some amino acids (d-Phe, l-/d-DOPA, d-Trp, l-Tyr) and amines (dopamine, serotonin, l-adrenaline and 4-(2-aminoethyl)-morpholine) with activation constants in the range of 4.0-9.5muM. l-/d-His, l-Phe, histamine and several other heterocyclic amines showed less efficient activation (K(A)s in the range of 11.6-33.4muM). The activation profile of CA XV is quite different from that of the cytosolic isoforms CA I and II or the membrane-associated CA IV. All mammalian isoforms CA I-XV are thus characterized for their interaction with this set of amino acid and amine activators, some of which are biogenic amines or neurotransmitters present in sufficiently high amounts in various tissues for exerting significant biologic responses.
Product Description
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
L-Histidine
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
1-(2-Aminoethyl)piperazine, 99%
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
D-Phenylalanine, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
D-Tryptophan, ≥98.0% (HPLC)
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
SAFC
L-Tyrosine
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
D-Histidine, ≥98% (TLC)
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
3,4-Dihydroxy-D-phenylalanine, powder, ≥95%
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
2-(2-Pyridyl)ethylamine, 95%
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
2-Picolylamine, 99%
Sigma-Aldrich
4-(2-Aminoethyl)morpholine, 99%