Skip to Content
Merck
All Photos(5)

Key Documents

P2126

Sigma-Aldrich

L-Phenylalanine

≥98%

Synonym(s):

(S)-2-Amino-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$142.00
500 G
$581.00
1 KG
$863.00

$142.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$142.00
500 G
$581.00
1 KG
$863.00

About This Item

Linear Formula:
C6H5CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
165.19
Beilstein:
1910408
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

$142.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

L-Phenylalanine, reagent grade, ≥98%

grade

reagent grade

Quality Level

Assay

≥98%

form

powder or crystals

color

white to off-white

mp

270-275 °C (dec.) (lit.)

solubility

1 M NH4OH: 50 mg/mL, clear, colorless

application(s)

detection

SMILES string

N[C@@H](Cc1ccccc1)C(O)=O

InChI

1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m0/s1

InChI key

COLNVLDHVKWLRT-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Phenylalanine has been used in Fluo-4 Ca2+-assay in a study to demonstrate that G-protein coupled receptor 139 reference surrogate agonists 1a and 7c and L-phenylalanine share a common binding site.[1] It has also been used to spike blood samples to study the effect of dried blood spots on newborn screening.[2]

Biochem/physiol Actions

L-Phenylalanine is an essential aromatic amino acid.[3][4] It has a role in metabolism as it forms a starting point for many pathways.[5] This neutral amino acid is a precursor for tyrosine, epinephrine, dopamine and norepinephrine.[5][5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein and Amino Acid Metabolism
ESSENTIALS OF MEDICINAL CHEMISTRY null
AMINO ACIDS | Metabolism
Encyclopedia of Food and Color Additives, 197-206 (2003)
Effect of Dried Blood Spot Quality on Newborn Screening Analyte Concentrations and Recommendations for Minimum Acceptance Criteria for Sample Analysis
Roanna S. George, Stuart J. Moat
Clinical Chemistry (2016)
The GPR139 reference agonists 1a and 7c, and tryptophan and phenylalanine share a common binding site
Anne Cathrine
Scientific Reports (2017)
Genetics in Medicine : Official Journal of the American College of Medical Genetics (2012)

Questions

1–4 of 4 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How long is Phenylalanine stable for when dissolved in water?

    1 answer
    1. The long term stability of this product in solution has not been investigated. However, this material is a component in several amino acid solutions that are stable for at least 1 year when filter sterilized and stored refrigerated at 2 - 8°C. Similar stability is expected with the individual component. Please see the link below to review the formulation sheet for these amino acid solutions:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/334/224/m7145for.pdf

      In addition, the stability information available for product 5202, L-Phenylalanine, indicates that this amino acid is stable for up to 1 year when stored at -20°C. Avoid repeated freeze/thaw cycles.

      Helpful?

  4. How long Phenylalanin can be stored as a powder at RT? Thank you. Erics

    1 answer
    1. This product may be stored at room temperature in the powder form until the recommended retest date published in the lot specific Certificate of Analysis. This compound is assigned a rested date of approximately 5 years from the date of Quality Release. Please see the link below to review a sample Certificate:
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/P2126-100G0000240439.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service