Skip to Content
Merck
All Photos(5)

Key Documents

D9628

Sigma-Aldrich

3,4-Dihydroxy-L-phenylalanine

≥98% (TLC)

Synonym(s):

3-(3,4-Dihydroxyphenyl)-L-alanine, L-3-Hydroxytyrosine, L-DOPA, Levodopa

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
197.19
Beilstein:
2215169
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.32

product name

3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)

Quality Level

Assay

≥98% (TLC)

form

powder

color

white to off-white

mp

276-278 °C (lit.)

application(s)

peptide synthesis

storage temp.

room temp

SMILES string

N[C@@H](Cc1ccc(O)c(O)c1)C(O)=O

InChI

1S/C9H11NO4/c10-6(9(13)14)3-5-1-2-7(11)8(12)4-5/h1-2,4,6,11-12H,3,10H2,(H,13,14)/t6-/m0/s1

InChI key

WTDRDQBEARUVNC-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,4-Dihydroxy-L-phenylalanine or L-DOPA has been used to stain melanocytes. It has also been used to study its effects on a Drosophila model of Parkinson′s disease.

Biochem/physiol Actions

3,4-Dihydroxy-L-phenylalanine or L-DOPA is a natural isomer of the immediate precursor of dopamine that crosses the blood-brain barrier. It is used for the treatment of Parkinson′s disease and is a product of tyrosine hydroxylase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of small-molecule amyloid modulators on a Drosophila model of Parkinson?s disease
Malgorzata Pokrzywa
PLoS ONE (2017)
Alan J. Sinclair, John E. Morley, Bruno Vellas
Pathy's Principles and Practice of Geriatric Medicine (2012)
Primary culture of human face skin melanocytes for the study of hyperpigmentation
Cytotechnology, 66(6) (2014)
Molly J Crockett et al.
Current biology : CB, 25(14), 1852-1859 (2015-07-07)
An aversion to harming others is a core component of human morality and is disturbed in antisocial behavior. Deficient harm aversion may underlie instrumental and reactive aggression, which both feature in psychopathy. Past work has highlighted monoaminergic influences on aggression
Sebastiaan P van Kessel et al.
Nature communications, 10(1), 310-310 (2019-01-20)
Human gut microbiota senses its environment and responds by releasing metabolites, some of which are key regulators of human health and disease. In this study, we characterize gut-associated bacteria in their ability to decarboxylate levodopa to dopamine via tyrosine decarboxylases.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service