Merck
  • Daily roadside BTEX concentrations in East Asia measured by the Lanwatsu, Radiello and Ultra I SKS passive samplers.
Daily roadside BTEX concentrations in East Asia measured by the Lanwatsu, Radiello and Ultra I SKS passive samplers.
The Science of the total environment (2012-11-13)
Tran Thi Ngoc Lan, Nguyen Thi Thanh Binh
ABSTRACT
A new home-made diffusive bag-type passive sampler called Lanwatsu was developed for benzene, toluene, ethylbenzene and xylene monitoring in roadside air. The passive samplers were outdoor validated and deployed together with two commercial passive samplers, Ultra I SKC Inc. and Radiello, for daily roadside air monitoring in East Asian cities including HoChiMinh, Hanoi, Cantho, Danang, Vungtau, Hue (Vietnam), Kuala Lumpur (Malaysia), Kyoto, Osaka (Japan), Nanjing (China) and Singapore in 2011. High daily benzene concentrations of 87, 52, 32, 23, 13, 12 and 48 µg/m³ were observed in HoChiMinh, Hanoi, Cantho, Danang, Hue, Vung Tau (Vietnam), and Kuala Lumpur (Malaysia), respectively. Kyoto and Osaka (Japan) were clean with daily benzene concentrations below 2.3 μg/m³. The daily benzene concentrations in Nanjing (China) and Singapore were 5.6 and 6.9 μg/m³, respectively. The three passive samplers were equivalent. Passive sampling by the Lanwatsu passive sampler is acceptable for daily outdoor benzene monitoring.
Product Description
radiello BTEX/VOC Starter Kit, Thermal Desorption
Supelco
radiello BTEX Calibration Kit (Thermal Desorption)
radiello Clips, suspending Radiello triangular support plate, pk of 20
radiello Filtration Kit, pk of 20
radiello On-Field Thermometer, Includes one standard Radiello vertical adapter (RAD122) for each thermometer, configured for standard use, pk of 3
radiello BTEX/VOC Starter Kit, CS2 Desorption
radiello Diffusive Bodies, white, configured for general use, pk of 20
radiello On-Field Thermometer, Reader, pk of 1
radiello Diffusive Bodies, blue, configured for sampling light sensitive compounds, pk of 20
Supelco
radiello BTEX Calibration Kit (CS2 Desorption)
radiello Empty Storage Tubes, glass (with stopper), volume 2.8 mL, pk of 100
radiello Diffusive Bodies, yellow, configured for reduced sampling rates, pk of 20
radiello Diffusive Bodies, permeative, configured for anaesthetic gases and vapors, pk of 20
Supelco
radiello Aldehyde Calibration Standard, 50 μg/mL each component in acetonitrile, pkg of 10 mL
radiello 1,3-Butadiene Starter Kit, Thermal Desorption, pkg of 1 kit (2 RAD141 adsorbing cartridges and barcode labels, 1 RAD1202 yellow diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet)
Supelco
radiello Methylene Blue Calibration Standard for H2S (Hydrogen Sulfide), pkg of 100 mL
radiello Empty Storage Tubes, polypropylene (with stopper), volume 12 mL, pk of 100
radiello Hydrogen Sulfide Starter Kit
radiello Cartridge Adsorbents, for sampling Anaesthetic Gases/Vapors, matrix SS net with mix of mol sieve and activated charcoal (30-50 mesh), pk of 20
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (thermal desorption), matrix SS net (3 x 8 μm, 4.8 mm diam.), Carbograph, pk of 20
radiello Triangular Support Plate, pk of 20
radiello Vertical Adapter, threaded for standard use, pk of 20
radiello Cartridge Adsorbents, for sampling Ammonia (NH3), matrix microporous PE impregnated with phosphoric acid, pk of 20
radiello Outdoor Shelter, pk of 10
radiello Replacement Mounting Strips, for use with Radiello Outdoor Shelter, pk of 100
radiello Cartridge Adsorbents, for sampling Aldehydes, matrix SS net with 2,4-DNPH coated FLORISIL®, pk of 20
radiello Barcode Labels, pk of 198
radiello Anesthetic Gases and Vapor Sampler
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (CS2 Desorption), matrix SS net (100 mesh, 5.8 mm diam.), activated with activated charcoal (30-50 mesh), pk of 20