Skip to Content
Merck
Phosphate buffered saline with 5% nonfat milk, powder (dry milled), pH 7.3

Phosphate buffered saline

Phosphate buffered saline with 5% nonfat milk, powder (dry milled), pH 7.3
Synonym(s):
PBS, Phosphate buffered saline

Products

Product Category
Brand
Color
Feature
pH Value
Physical Form
Product Line
Research Area
Chemical Composition
Quality Segment
Application
Sterilization Method
Technique
Showing 1-20 of 22
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2