Skip to Content
Merck
All Photos(1)

Key Documents

P2194

Sigma-Aldrich

Phosphate Buffered Saline

powder, pH 6.9-7.9 ( in water, 1 pouch dissolved in 100 ml), with 3% Non-Fat Milk

Synonym(s):

NFDM blocking buffer, PBS with 3% NFDM, Phosphate Buffered Saline with 3% Non-Fat Dry Milk

Sign Into View Organizational & Contract Pricing

Select a Size

10 PKG
$305.00
5 X 10 PKG
$1,220.00

$305.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability
Diagnostics Manufacturers should use alternative grade product: SRE0036

Request a Bulk Order

Select a Size

Change View
10 PKG
$305.00
5 X 10 PKG
$1,220.00

About This Item

UNSPSC Code:
41105319
eCl@ss:
32129211
NACRES:
NA.25

$305.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability
Diagnostics Manufacturers should use alternative grade product: SRE0036

Request a Bulk Order

Product Name

Phosphate Buffered Saline with 3% Non-Fat Milk, pH 7.4,

form

powder

pH

6.9-7.9 ( in water, 1 pouch dissolved in 100 ml)

solubility

water: soluble

Looking for similar products? Visit Product Comparison Guide

Application

Phosphate Buffered Saline with 3% Non-Fat Milk, pH 7.4 has been used as a resuspension buffer during umbilical cord perivascular cells separation.[1]

Packaging

Foil pouches

Reconstitution

Each pouch dissolved in 100 mL deionized water will yield 0.05 M phosphate buffered saline, pH 7.4 at 25 °C, containing 3% (w/v) nonfat milk.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rodrigo Viana Sepúlveda et al.
Research in veterinary science, 129, 193-202 (2020-02-23)
There are numerous sources of multipotent mesenchymal stromal cells (MSC) with therapeutic potential, and bone marrow is the main one. However, pain, lack of donors and comorbidities associated with harvesting stimulate the search for new sources of MSCs. The aim
Sensitivity of Vegetative Pathogens to High Hydrostatic Pressure Treatment in Phosphate-Buffered Saline and Foods
Patterson MF, et al.
Journal of Food Protection (1995)
Konrad Franke et al.
Translational oncology, 2(4), 310-320 (2009-12-04)
Nodal spread is the single most important prognostic factor of survival in gastric cancer patients. In this study, genes that were upregulated in the lymph node metastases of gastric cancer were identified and may serve as putative novel therapeutic target.
Yong N Li et al.
The Journal of comparative neurology, 516(5), 442-453 (2009-08-06)
Horizontal cells (HCs) are involved in establishing the center-surround receptive field organization of photoreceptor and bipolar cells. In many species, HCs respond differentially to colors and may play a role in color vision. An earlier study from our laboratory suggested
Campylobacter jejuni in penguins, Antarctica.
Petra Griekspoor et al.
Emerging infectious diseases, 15(5), 847-848 (2009-05-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service