Skip to Content
Merck

Skip To

PPB006

Phosphate buffered saline

powder, pH 7.4, pHast Pack

Synonym(s):

1X PBS wash buffer, PBS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 pkg
Check Cart for Availability
$137.00
$116.45

About This Item

UNSPSC Code:
12161700
NACRES:
NA.21
MDL number:

$116.45

List Price$137.00Save 15%
PRICE WITHOUT NATIONAL TAXES
Web-Only Promotion

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Phosphate buffered saline, pH 7.4, pHast Pack, powder

product line

pHast Pack

form

powder

storage condition

(Tightly closed. Dry)

impurities

<5—2 ppm Heavy metals (Lead)

color

white

pH

7.4

solubility

soluble, clear, colorless

cation traces

Fe: <10 ppm

foreign activity

DNAse, none detected, Nickase, none detected, Protease, none detected, RNAse, none detected

storage temp.

room temp

General description

Phosphate buffered saline or PBS, pH 7.4 is a commonly used biological buffer in molecular biology. A 1X working solution is typically used in cell culture and electrophoresis assays to wash away non-adherent cells or any unbound biomolecules. PBS is hypo-osmotic and can be harmful to cells, causing swelling and resulting in cell lysis, if left for extended periods.

Application

Phosphate buffered saline is commonly used in washing solutions for:
  • ELISA
  • Western Blotting and other protein applications
  • Cell culture (sterile filtration recommended)
  • Resuspension and sonication of cells
  • Immunohistochemistry

Features and Benefits

  • Save time with our ready-to-use formulation
  • No calculations, measurements, and adjusting pH
  • Reliable quality for reproducibility
  • Suitable for use in cell culture and electrophoresis
  • Biological tests: free of DNase, RNase, Protease, and Nickase
  • Chemical tests: Iron ≤10 ppm, Lead ≤5 ppm

Packaging

Foil pouches

Preparation Note

Contents of one pouch, when dissolved in 500mL of distilled or deionized water, will yield a 1X solution containing 10mM Na2HPO4, 1.8mM KH2PO4, 138mM NaCl, 2.7mM KCl, pH 7.4 at 25 °C. Certified to be DNAse, RNAse, Protease, and Nickase free. Certified for use in cell culture.
Store at room temperature. Product may naturally agglomerate but can be simply broken up within the pouch prior to use.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

pHast Pack is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
79378P5119P2272
product line

pHast Pack

product line

BioUltra

product line

-

product line

-

form

powder

form

solution

form

liquid

form

liquid

solubility

soluble, clear, colorless

solubility

-

solubility

-

solubility

-

storage condition

(Tightly closed. Dry)

storage condition

-

storage condition

-

storage condition

-

storage temp.

room temp

storage temp.

-

storage temp.

2-8°C

storage temp.

-

pH

7.4

pH

7.1-7.3

pH

7.2 (25 °C)

pH

7.2


Still not finding the right product?

Explore all of our products under Phosphate buffered saline


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. What is the weight and dimensions of the packaging?

    1 answer
    1. The weight of the packaging is 160 grams.

      Regarding the dimensions, We kindly ask you to navigate to the link https://www.sigmaaldrich.com/techservice, click on "Product Documentation" under the Products Section with all the required information so that a member of our team can reach out to you to assist further. Thank you.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters