Bioscience, biotechnology, and biochemistry, 66(11), 2388-2394 (2003-01-01)
After enrichment culture with indole-3-carboxylate in static culture, a novel reversible decarboxylase, indole-3-carboxylate decarboxylase, was found in Arthrobacter nicotianae FI1612 and several molds. The enzyme reaction was examined in resting-cell reactions with A. nicotianae FI1612. The enzyme activity was induced
Journal of the National Cancer Institute, 82(11), 947-949 (1990-06-06)
Dietary indoles in cruciferous vegetables induce cytochrome P450 enzymes and have prevented tumors in various animal models. Because estradiol metabolism is also cytochrome P450 mediated and linked to breast cancer risk, indoles may similarly reduce estrogen-responsive tumors in humans. We
Current opinion in plant biology, 15(4), 407-414 (2012-03-27)
In plants, a host's responses to an attempted infection include activation of various secondary metabolite pathways, some of which are specific for particular plant phylogenetic clades. Phytochemicals that represent respective end products in plant immunity have been stereotypically linked to
The Journal of organic chemistry, 77(15), 6505-6509 (2012-07-19)
Elucidation of the mechanism for decarboxylation of indolecarboxylic acids over a wide range of solution acidity reveals the importance of protonated carbonic acid (PCA) as a reaction intermediate. In concentrated acid, the initial addition of water to the carboxyl group
The Journal of organic chemistry, 72(21), 8115-8118 (2007-09-20)
An enantioselective synthetic method for (-)-cis-clavicipitic acid (1) was reported. 1 was obtained in 10 steps (99% ee and 20% overall yield) from 1H-indole-3-carboxylic acid methyl ester (9) via asymmetric phase-transfer catalytic alkylation and diastereoselective Pd(II)-catalyzed intramolecular aminocyclization as key
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.