Skip to Content
Merck
All Photos(1)

Key Documents

809780

Sigma-Aldrich

Indoxyl-3a,4,5,6,7,7a-13C6 sulfate potassium salt

≥99 atom % 13C, ≥95% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,040.00

$1,040.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request more information

Select a Size

Change View
1 MG
$1,040.00

About This Item

Empirical Formula (Hill Notation):
13C6C2H6KNO4S
Molecular Weight:
257.26
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

$1,040.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request more information

isotopic purity

≥99 atom % 13C

Quality Level

Assay

≥95% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+6

storage temp.

−20°C

SMILES string

O=S(C1=CN[13C]2=[13CH][13CH]=[13CH][13CH]=[13C]21)([O-])=O.[K+]

InChI

1S/C8H7NO3S.K/c10-13(11,12)8-5-9-7-4-2-1-3-6(7)8;/h1-5,9H,(H,10,11,12);/q;+1/p-1/i1+1,2+1,3+1,4+1,6+1,7+1;

InChI key

KOCDDYKTQUAJNN-BTZXTVEYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service