V900176
Iron(III) sulfate hydrate
Vetec™, reagent grade, 97%
Synonym(s):
Ferric sulfate hydrate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
Fe2(SO4)3 · xH2O
CAS Number:
Molecular Weight:
399.88 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
Assay:
97%
grade:
reagent grade
form:
powder
Recommended Products
grade
reagent grade
product line
Vetec™
Assay
97%
form
powder
reaction suitability
reagent type: catalyst
core: iron
SMILES string
[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
InChI
1S/2Fe.3H2O4S/c;;3*1-5(2,3)4/h;;3*(H2,1,2,3,4)/q2*+3;;;/p-6
InChI key
RUTXIHLAWFEWGM-UHFFFAOYSA-H
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M E Odabaş et al.
European journal of paediatric dentistry : official journal of European Academy of Paediatric Dentistry, 13(2), 118-122 (2012-07-06)
The purpose of this study was to evaluate and compare the clinical and radiographic findings of ferric sulphate (FS) and mineral trioxide aggregate (MTA) as vital pulpotomy materials in primary molars. Participants, including 40 boys and 53 girls ranging between
M I Murav'ev et al.
Prikladnaia biokhimiia i mikrobiologiia, 47(6), 663-671 (2012-02-01)
Leaching of sulfide-oxidized copper concentrate of the Udokan deposit ore with a copper content of 37.4% was studied. In the course of treatment in a sulfuric acid solution with pH 1.2, a copper leaching rate was 6.9 g/kg h for
H Y Zhang et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 64(7), 1413-1418 (2011-12-20)
This work aims to investigate removal efficiency of oxidation and coagulation/flocculation processes, to provide an effective method for the treatment of biologically pre-treated leachate. Leachate containing 985 mg L(-1) COD was treated by using three treatment schemes, i.e. oxidation, coagulation/flocculation
Gal Frenkel et al.
The Journal of clinical pediatric dentistry, 37(2), 137-141 (2012-01-01)
To compare the long-term clinical and radiographic outcomes of pulpotomies in primary molars performed with white or gray Mineral Trioxide Aggregate (MTA) in combination with ferric sulfate (FS), when one package of MTA is used for multiple treatments. Sixty eight
Y Yilmaz et al.
European journal of paediatric dentistry : official journal of European Academy of Paediatric Dentistry, 12(2), 117-122 (2011-06-15)
The aim of this study was to compare the local anaesthetic efficacy of articaine HCl and prilocaine HCl during an operative procedure after their administration by either mandibular nerve block or maxillary infiltration. The study was a double-blind clinical study
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service