Skip to Content
Merck
All Photos(4)

Key Documents

217255

Sigma-Aldrich

Potassium sodium tartrate tetrahydrate

ACS reagent, 99%

Synonym(s):

L(+)-Tartaric acid potassium Sodium salt, Rochelle salt, Seignette salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$152.00
500 G
$399.00
2.5 KG
$1,120.00
12 KG
$2,240.00

$152.00


Estimated to ship onMarch 30, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$152.00
500 G
$399.00
2.5 KG
$1,120.00
12 KG
$2,240.00

About This Item

Linear Formula:
KOCOCH(OH)CH(OH)COONa · 4H2O
CAS Number:
Molecular Weight:
282.22
Beilstein:
6113568
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
99%
99.0-102.0% (ACS specification)
grade:
ACS reagent
form:
powder or crystals

$152.00


Estimated to ship onMarch 30, 2025


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

suitable for SM 4500 - NH3

Assay

99%
99.0-102.0% (ACS specification)

form

powder or crystals

impurities

≤0.005% insolubles

pH

6.0-8.5

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.002%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.005%
Fe: ≤0.001%
NH4+: ≤0.002%
heavy metals: ≤5 ppm (by ICP)

SMILES string

O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

InChI

1S/C4H6O6.K.Na.4H2O/c5-1(3(7)8)2(6)4(9)10;;;;;;/h1-2,5-6H,(H,7,8)(H,9,10);;;4*1H2/q;2*+1;;;;/p-2/t1-,2-;;;;;;/m1....../s1

InChI key

VZOPRCCTKLAGPN-ZFJVMAEJSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Potassium sodium tartrate tetrahydrate is a ferroelectric crystal[1] with a high piezoelectric effect and electromechanical coupling coefficient.[2]

Application

Potassium sodium tartrate tetrahydrate has been used in the preparation of Lowry reagent for the determination of microsomal protein concentration in rat hepatic microsomes by Lowry method.[3] It may be used as a constituent to prepare DNS (3,5- dinitrosalicylic acid) reagent and Fehling′s solution B, which are used in the determination of reducing sugar.[4][5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Organic nonvolatile memory devices based on ferroelectricity.
Naber RCG, et al.
Advanced Materials, 22(9), 933-945 (2010)
Fundamentals of Piezoelectric Sensorics: Mechanical, Dielectric, and Thermodynamical Properties of Piezoelectric Materials, 11-11 (2010)
In vitro Studies: Inhibition of Nevirapine Metabolism by Nortriptyline in Hepatic Microsomes
Usach I and Peris JE.
Antimicrobial Agents and Chemotherapy (2014)
Screening of African traditional vegetables for their alpha-amylase inhibitory effect.
Odhav B, et al.
Journal of Medicinal Plants Research, 4(14), 1502-1507 (2010)
Wen-Bin Du et al.
Analytical chemistry, 82(23), 9941-9947 (2010-11-04)
This paper describes DropLab, an automated microfluidic platform for programming droplet-based reactions and screening in the nanoliter range. DropLab can meter liquids with picoliter-scale precision, mix multiple components sequentially to assemble composite droplets, and perform screening reactions and assays in

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service