Skip to Content
Merck
All Photos(1)

Key Documents

PHR1248

Supelco

Maltitol

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

4-O-α-Glucopyranosyl-D-sorbitol

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$177.00

$177.00


Estimated to ship onMarch 25, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$177.00

About This Item

Empirical Formula (Hill Notation):
C12H24O11
CAS Number:
Molecular Weight:
344.31
Beilstein:
89983
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$177.00


Estimated to ship onMarch 25, 2025


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. M0160000
traceable to USP 1374907

API family

maltitol

CofA

current certificate can be downloaded

analyte chemical class(es)

oligosaccharides

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

149-152 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

OC[C@H](O)[C@@H](O)[C@H](O[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H](O)CO

InChI

1S/C12H24O11/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12/h4-21H,1-3H2/t4-,5+,6+,7+,8+,9-,10+,11+,12+/m0/s1

InChI key

VQHSOMBJVWLPSR-WUJBLJFYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Maltitol is a sugar alcohol and nutritive sweetener, exhibiting reduced laxative effects,[1] hence finds applications in different food products, beverages and dietary supplements.[2]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Sugar substitute with a weak genotoxic effect.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB7670 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of maltitol, isomaltitol, and lactitol by high-pH anion-exchange chromatography with pulsed amperometric detection
Cataldi.I.RT, et al.
Journal of Agricultural and Food Chemistry, 47(1), 157-163 (1999)
Simultaneous separation and determination of erythritol, xylitol, sorbitol, mannitol, maltitol, fructose, glucose, sucrose and maltose in food products by high performance liquid chromatography coupled to charged aerosol detector
Grembecka M, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 117, 77-82 (2014)
Pernilla Lif Holgerson et al.
International journal of paediatric dentistry, 17(2), 79-85 (2007-02-01)
The aim of this study was to investigate the effect of a fixed daily dose of xylitol on mutans streptococci in saliva and the amount of visible dental plaque. A second aim was to explore if the possible effects differed
Aija-Maaria Hietala Lenkkeri et al.
International journal of paediatric dentistry, 22(3), 180-190 (2011-09-29)
Xylitol studies suggest caries reductions in the order of 50%. Based on animal/microbial studies, erythritol potentially has caries-preventive properties. However, clinical studies are required to confirm this. The aim of the study was to investigate the additional caries-preventive effect of
Xiaoming Liu et al.
Journal of agricultural and food chemistry, 57(6), 2339-2345 (2009-02-24)
The objective of this study was to investigate the influence of polyols on the stability of whey proteins in an intermediate-moisture food model system and to elucidate the effect of polyols on the hardening of whey protein-based bars during storage.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service