Skip to Content
Merck
All Photos(1)

Key Documents

CRM46912

Supelco

Ochratoxin A solution

certified reference material, 50 μg/mL in benzene: acetic acid (99:1), ampule of 1 mL

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$175.00

$175.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 23, 2025Details


Request a Bulk Order

Select a Size

Change View
1 ML
$175.00

About This Item

Empirical Formula (Hill Notation):
C20H18ClNO6
CAS Number:
Molecular Weight:
403.81
Beilstein:
8169012
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

$175.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 23, 2025Details


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

50 μg/mL in benzene: acetic acid (99:1)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

-10 to -25°C

SMILES string

C[C@@H]1Cc2c(Cl)cc(c(O)c2C(=O)O1)C(=O)N[C@@H](Cc3ccccc3)C(O)=O

InChI

1S/C20H18ClNO6/c1-10-7-12-14(21)9-13(17(23)16(12)20(27)28-10)18(24)22-15(19(25)26)8-11-5-3-2-4-6-11/h2-6,9-10,15,23H,7-8H2,1H3,(H,22,24)(H,25,26)/t10-,15+/m1/s1

InChI key

RWQKHEORZBHNRI-BMIGLBTASA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Ochratoxin A, a mycotoxin, is a naturally occurring secondary metabolite, produced by filamentous fungi, such as Aspergillus and Penicillium. It has been classified as a group 2B substance by International Agency for Research on Cancer, for possibly being carcinogenic to humans.

Application

The CRM can also be used as follows:
  • Identification and determination of ochratoxin A in different types of commercial coffee samples using dispersive liquid-liquid microextraction-solidification of a floating organic drop (DLLME-SFO) for sample treatment and ultra high-performance liquid chromatography-positive electrospray ionization-tandem mass spectrometry (UHPLC-(+-ESI)-MS/MS)
  • Determination of ochratoxin A in beer samples by a selective method based on salting-out assisted liquid-liquid extraction (SALLE) and ultra high-performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)
  • Simultaneous determination of ochratoxin A and ochratoxin B in Tokaj wine samples using online solid phase extraction (SPE) combined with high-performance liquid chromatography-fluorescence detection(HPLC-FD)
  • Development of an online-SPE-HPLC method to determine ochratoxin A in wine samples
  • Electrochemical determination of ochratoxin A using an aptamer-based on functionalized graphene (f-graphene) doped chitosan in grape juice samples by differential pulse voltammetry

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cannabis testing regulations include analysis of purity, potency, and contaminants. Learn more about analysis of cannabis for pesticides, mycotoxins, and pyrethrins registered in California by LC-MS/MS and GC-MS/MS Using Ascentis® Express Biphenyl and SLB®-5ms Columns

Related Content

Detailed list of mycotoxin analysis products for Food & Beverage manufacturers and testers, ensuring safety standards.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service