Skip to Content
Merck
All Photos(1)

Key Documents

BCR471

Wheat (ochratoxin A, blank)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

55 G
$177.00

$177.00


Estimated to ship onApril 09, 2025


Request a Bulk Order

Select a Size

Change View
55 G
$177.00

About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

$177.00


Estimated to ship onApril 09, 2025


Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

format

matrix material

storage temp.

2-8°C

Analysis Note

For more information please see:
BCR471

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Biotoxins / Mycotoxins: Ochratoxin A

Matrix Group: Cereals, Bread, Rice and Maize

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral - Carc. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service