Skip to Content
Merck
All Photos(1)

Key Documents

CRM46324

Supelco

Aflatoxin B2 solution

certified reference material, 3 μg/mL in benzene:acetonitrile (98:2), ampule of 1 mL

Synonym(s):

Aflatoxin B2 solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$111.00

$111.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$111.00

About This Item

Empirical Formula (Hill Notation):
C17H14O6
CAS Number:
Molecular Weight:
314.29
Beilstein:
1355115
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$111.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

3 μg/mL in benzene:acetonitrile (98:2)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

[H][C@@]12CCO[C@]1([H])Oc3cc(OC)c4C5=C(C(=O)CC5)C(=O)Oc4c23

InChI

1S/C17H14O6/c1-20-10-6-11-14(8-4-5-21-17(8)22-11)15-13(10)7-2-3-9(18)12(7)16(19)23-15/h6,8,17H,2-5H2,1H3/t8-,17+/m0/s1

InChI key

WWSYXEZEXMQWHT-WNWIJWBNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cormac McElhinney et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 32(12), 2101-2112 (2015-09-17)
An ultra-high-performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) multi-mycotoxin analytical method was developed to simultaneously identify and quantify 20 mycotoxins in grass silages, inclusive of mycotoxins that are currently regulated in European Union feeds. Extraction of mycotoxins from dried grass silages
Emmanuel Njumbe Ediage et al.
Food chemistry, 177, 397-404 (2015-02-11)
An LC-MS/MS method was developed and validated for the detection and quantification of 23 mycotoxins in different varieties of sorghum. The method performance characteristics were as follows: suitable linearity ranges for all 23 mycotoxins with p-value >0.05; limits of detection

Protocols

LC/MS/MS Analysis of Aflatoxins in Hops on Ascentis® Express 2.7 μm Phenyl-Hexyl after Cleanup Using Supel™ Tox AflaZea SPE

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service