Skip to Content
Merck
All Photos(1)

Documents

814701

TOYOPEARL® Butyl-650S Bulk Media

phase C4 (butyl), bottle of 1 L, 35 μm particle size

Synonym(s):

TOYOPEARL® Butyl-650 HIC Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

description

Butyl-650S

product line

TOYOPEARL®

form

slurry

packaging

bottle of 1 L

manufacturer/tradename

Tosoh 14701

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

C4 (butyl) phase

particle size

35 μm

operating pH

1-13

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

TOYOPEARL® HIC resins are hydrophobic interaction chromatography resins that offer the following advantages: strong affinity for water-soluble proteins, high recovery of mass and activity, high sample capacity up to 2-4 times that of gel media, fluctuating salt concentrations will not change the bed volume, mechanical stability to 7kg/cm squared (7 bar/100psi), stability at a wide pH range (2-12), clean in place with 0.5M NaOH, and autoclavable. The exclusion limit of 5x106 Da and large pore size, 1000Å, enables these packings to separate very large proteins by a hydrophobic interaction mechanism, without size exclusion effects.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Other Notes

Toyopearl Butyl-650 resin has the second highest hydrophobicity of the HIC ligands offered by TBL. Its pore size is the largest of our three butyl resins. Primary applications are for the separation of proteins, their isoforms, and aggregate removal. It is typically used in interresinte purification and polishing steps

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service