Key Documents
Z614009
Sigma-Aldrich
Vivaspin 500 centrifugal concentrators
MWCO 5 kDa
Synonym(s):
Vivaspin 500
Select a Size
25 EA
$238.00
$238.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
25 EA
$238.00
About This Item
UNSPSC Code:
41104916
NACRES:
NB.24
$238.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
polycarbonate housing
polyethersulfone membrane
description
PES membrane
sterility
non-sterile
feature
holdup volume 5 μL
packaging
package of 25 ea
manufacturer/tradename
Sartorius VS0111
parameter
15000 × g max. centrifugal force
dead stop volume
5 μL
membrane area
0.5 cm2
tube size
~2.2 mL
working volume
500 μL
pore size
5 kDa MWCO
General description
  • Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs.
  • Highest flow rates and recoveries are achieved due to their particular vertical membrane design.
  • Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin.
Note: Vivaspin 500 μL centrifugal filter units offer a simple one step procedure for sample preparation. They can effectively be used in either swing out or fixed angle rotors.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Przemyslaw A Filipek et al.
The Journal of cell biology, 216(12), 4199-4215 (2017-10-11)
Signaling from lysosomes controls cellular clearance and energy metabolism. Lysosomal malfunction has been implicated in several pathologies, including neurodegeneration, cancer, infection, immunodeficiency, and obesity. Interestingly, many functions are dependent on the organelle position. Lysosomal motility requires the integration of extracellular
Questions
  1. can i concentrate a working volume of 100 microliters (ul)?
    1 answer
    1. Yes, the maximum capacity is 500 µL. This product is manufactered by Sartorius under the product number VS0111. For more information please view manufacturer webpage: https://www.sartorius.com/shop/ww/applications-laboratory-filtration-ultrafiltration/vivaspin-500%2C-5%2C000-mwco-pes%2C-25pc/p/VS0111
      Helpful?