Skip to Content
Merck
All Photos(7)

Key Documents

323136

Sigma-Aldrich

Trimethylboroxine

99%

Synonym(s):

2,4,6-Trimethyl-1,3,5,2,4,6-trioxatriborinane, 2,4,6-Trimethylboroxine, Methaneboronic anhydride, Trimethyl-1,3,5,2,4,6-trioxatriborinane

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$67.40
5 G
$170.00
25 G
$444.00
250 G
$3,380.00

$67.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$67.40
5 G
$170.00
25 G
$444.00
250 G
$3,380.00

About This Item

Empirical Formula (Hill Notation):
C3H9B3O3
CAS Number:
Molecular Weight:
125.53
Beilstein:
1757008
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$67.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.362 (lit.)

bp

78-80 °C (lit.)

mp

−38 °C (lit.)

density

0.898 g/mL at 25 °C (lit.)

SMILES string

Cb1ob(C)ob(C)o1

InChI

1S/C3H9B3O3/c1-4-7-5(2)9-6(3)8-4/h1-3H3

InChI key

GBBSAMQTQCPOBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trimethylboroxine (TMB) is a cyclic anhydride of methyl-boronic acid. It can be been used:
  • As a derivatizing agent for gas chromatographic/mass spectrometric analysis.[1]
  • In the preparation of methylaluminoxane (MAO) which is used in the polymerization of olefins.[2]
  • As an electrolyte additive to enhance the interface stability of electrode/electrolyte.[3]
  • In methylation of aryl halides by palladium-catalyzed Suzuki-Miyaura coupling reaction.[4]
  • In the preparation of CBS (Corey, Bakshi and Shibata) catalysts for asymmetric reductions of ketones to alcohols.[5]

related product

Product No.
Description
Pricing

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

15.8 °F - closed cup

Flash Point(C)

-9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymer, 37, 4629-4629 (1996)
Enhanced high voltage performances of layered lithium nickel cobalt manganese oxide cathode by using trimethylboroxine as electrolyte additive
Yu Q, et al.
Electrochimica Acta, 176(14), 919-925 (2015)
Reactivity of metallocene catalysts for olefin polymerization: influence of activator nature and structure
Pedeutour JN
Macromolecular Rapid Communications, 22(14), 1095-1123 (2001)
Tetrahedron Asymmetry, 14, 2115-2118 (2003)
Practical methylation of aryl halides by Suzuki-Miyaura coupling.
Gray M, et al.
Tetrahedron Letters, 41(32), 6237-6240 (2000)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service