Skip to Content
Merck
All Photos(2)

Key Documents

44892-U

Supelco

ASTM® D6584 Glycerin solution

500 μg/mL in pyridine, analytical standard

Synonym(s):

Glycerol solution, Biodiesel impurity, 1,2,3-Propanetriol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH · aq
CAS Number:
Molecular Weight:
92.09 (anhydrous basis)
Beilstein:
635685
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Agency

ASTM® D6584

packaging

pkg of 1 mL

concentration

500 μg/mL in pyridine

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

182 °C/20 mmHg (lit.)

density

1.252 g/mL at 25 °C (lit.)

application(s)

food and beverages
petroleum

format

single component solution

storage temp.

2-8°C

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jonathan S Earl et al.
American journal of dentistry, 26 Spec No A, 19A-24A (2013-07-10)
To characterize, in vitro, the mode of action of stannous fluoride containing formulations in occluding dentin tubules, by means of high resolution microscopy techniques. Focused ion beam scanning electron microscopy (FIB SEM) was used to site-specifically prepare cross sections for
Pavan K Ghattyvenkatakrishna et al.
Cryo letters, 34(2), 166-173 (2013-04-30)
The effect of glycerol, water and glycerol-water binary mixtures on the structure and dynamics of biomolecules has been well studied. However, a lot remains to be learned about the effect of varying glycerol concentration and temperature on the dynamics of
K O Tokarchuk et al.
Fiziolohichnyi zhurnal (Kiev, Ukraine : 1994), 59(1), 25-31 (2013-05-30)
We investigated the changes in the forms of plasma iron and participation of aldehydes in the development of oxidative stress under glycerol-induced rhabdomyolysis in rats. Rhabdomyolysis was caused by intramuscular injection of 50% glycerol in the dose 10 ml/kg. We
T M Hörer et al.
European journal of vascular and endovascular surgery : the official journal of the European Society for Vascular Surgery, 45(6), 596-606 (2013-04-02)
This study aims to evaluate intra-peritoneal (ip) microdialysis after endovascular aortic repair (EVAR) of ruptured abdominal aortic aneurysm (rAAA) in patients developing intra-abdominal hypertension (IAH), requiring abdominal decompression. Prospective study. A total of 16 patients with rAAA treated with an
L Maes et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 67(12), 2863-2867 (2013-06-22)
Sewage sludge and crude glycerine were co-digested in the mesophilic digester of Hoogstraten wastewater treatment plant. Additions of up to 1 kg of crude glycerine/(m³ reactor).(day) were done without significant operational problems. At higher dosage, severe digester foaming was observed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service