Merck
  • Effect of temperature and glycerol on the hydrogen-bond dynamics of water.
Effect of temperature and glycerol on the hydrogen-bond dynamics of water.
Cryo letters (2013-04-30)
Pavan K Ghattyvenkatakrishna, Edward C Uberbacher
ABSTRACT
The effect of glycerol, water and glycerol-water binary mixtures on the structure and dynamics of biomolecules has been well studied. However, a lot remains to be learned about the effect of varying glycerol concentration and temperature on the dynamics of water. We have studied the effect of concentration and temperature on the hydrogen bonded network formed by water molecules. A strong correlation between the relaxation time of the network and average number of hydrogen bonds per water molecules was found. The radial distribution function of water oxygen and hydrogen atoms clarifies the effect of concentration on the structure and clustering of water.
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)