Merck
  • Intra-peritoneal microdialysis and intra-abdominal pressure after endovascular repair of ruptured aortic aneurysms.
Intra-peritoneal microdialysis and intra-abdominal pressure after endovascular repair of ruptured aortic aneurysms.
European journal of vascular and endovascular surgery : the official journal of the European Society for Vascular Surgery (2013-04-02)
T M Hörer, P Skoog, L Norgren, A Magnuson, L Berggren, K Jansson, T Larzon
ABSTRACT
This study aims to evaluate intra-peritoneal (ip) microdialysis after endovascular aortic repair (EVAR) of ruptured abdominal aortic aneurysm (rAAA) in patients developing intra-abdominal hypertension (IAH), requiring abdominal decompression. Prospective study. A total of 16 patients with rAAA treated with an emergency EVAR were followed up hourly for intra-abdominal pressure (IAP), urine production and ip lactate, pyruvate, glycerol and glucose by microdialysis, analysed only at the end of the study. Abdominal decompression was performed on clinical criteria, and decompressed (D) and non-decompressed (ND) patients were compared. The ip lactate/pyruvate (l/p) ratio was higher in the D group than in the ND group during the first five postoperative hours (mean 20 vs. 12), p = 0.005 and at 1 h prior to decompression compared to the fifth hour in the ND group (24 vs. 13), p = 0.016. Glycerol levels were higher in the D group during the first postoperative hours (mean 274.6 vs. 121.7 μM), p = 0.022. The IAP was higher only at 1 h prior to decompression in the D group compared to the ND group at the fifth hour (mean 19 vs. 14 mmHg). Ip l/p ratio and glycerol levels are elevated immediately postoperatively in patients developing IAH leading to organ failure and subsequent abdominal decompression.
Sigma-Aldrich
Pyruvic acid, ≥97%, FG
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Pyruvic acid, 98%
Sigma-Aldrich
Pyruvic acid, 95%
Sigma-Aldrich
Pyruvic acid, natural, ≥95%, FG
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%