Skip to Content
Merck
All Photos(1)

Key Documents

21287-U

Supelco

GC Stationary Phase

phase SP-2330, bottle of 5 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.54

Agency

meets requirements for USP G8

form

solid

packaging

bottle of 5 g

parameter

25-275 °C temp. range

technique(s)

gas chromatography (GC): suitable

matrix active group

SP-2330 phase

solubility

acetone: soluble

column type

packed GC

General description

GC methods are divided into two classes depending on the nature of stationary phases; gas-solid chromatography (GSC) and gas-liquid chromatography (GLC). GSC has solid adsorptive material and solute particles are removed from mobile phase by electrostatic forces. GLC has a thin layer of liquid coated or bonded on the surface of an inert particle or on the walls of the column where solute particles are retained in the liquid phase based on their partition coefficients. The primary necessity of a stationary phase is to provide sample separation sustaining phase integrity over a reasonable period of time. It should be stable for the chemical and thermal changes. Selectivity, peak symmetry, analysis time, degree of separation, peak tailing are few parameters to consider for choosing a stationary phase. SP-2330 is a cyanosilicone suitable for individual isomer determination. It provides better separation from interfering components specially phthalate plasticizers.
Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.

Application

Capillary columns filled with SP-2330 is suitable in separating methyl esters of FA analysis suspended in methylene chloride.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D G Birch et al.
Investigative ophthalmology & visual science, 33(8), 2365-2376 (1992-07-01)
Full-field electroretinograms (ERGs) were obtained from very-low-birth-weight (VLBW) neonates to determine whether omega-3 (omega-3) fatty acids are essential for normal human retinal development. Eighty-one infants born at 30.4 (standard deviation, +/- 1.5) wk gestation were, within 10 d of birth
Gunter Zweig, Joseph Sherma
Synthetic Pyrethroids and Other Pesticides: Analytical Methods for Pesticides and Plant Growth Regulators, 118-118 (2013)
James P. Lodge, Jr.
Methods of Air Sampling and Analysis, 98-98 (1988)
David B. Troy, Paul Beringer
Remington: The Science and Practice of Pharmacy, 605-605 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service