Skip to Content
Merck
All Photos(3)

Documents

21024-U

Supelco

LpDNPH S10 solid phase extraction (SPE) Cartridge

Starter Kit, bed wt. 350 mg, volume 3 mL, pkg of (kit includes 10 tubes, 10 tube adapters, 10 male luer ⅛" fittings)

Synonym(s):

LpDNPH S10 Cartridge Tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000
NACRES:
NB.24

product name

LpDNPH S10 Cartridge, Starter Kit, bed wt. 350 mg, volume 3 mL, pkg of (kit includes 10 tubes, 10 tube adapters, 10 male luer ⅛" fittings)

Agency

ASTM® D5197
EPA 100,IP-6A,TO-11A,TO-11A
NIOSH 2016

composition

bed wt., 350 mg
2,4-DNPH (High-purity silica gel)

packaging

pkg of (kit includes 10 tubes, 10 tube adapters, 10 male luer ⅛" fittings)

technique(s)

active air sampling: suitable

volume

3 mL

application(s)

air monitoring
environmental
industrial hygiene

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Starter Kit includes: 10 cartridges, 1 tube adaptor, and 10 1/8 in. male luer fittings
The design of the S10 cartridge makes it easy to use in the field and laboratory. Reusable adaptors are available for connecting the cartridge to the sampling pump. The built in reservoir eliminates the need to attach a syringe for sample extraction/elution.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Kit Components Also Available Separately

Product No.
Description
SDS

  • 21026-ULpDNPH S10 Cartridge, bed wt. 350 mg, volume 3 mL, pkg of 10 ea 1SDS

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefano Dugheri et al.
Arhiv za higijenu rada i toksikologiju, 71(3), 178-189 (2020-10-20)
The last two decades have been crucial for the assessment of airborne formaldehyde (FA) exposure in healthcare environments due to changes in limits and reference values, definition of carcinogenicity, and new monitoring methods. The aim of this study was to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service