54072-U
LpDNPH S10 solid phase extraction (SPE) Cartridge
bed wt. 350 mg, volume 3 mL, pkg of 50 ea (1 per nylon zip bag)
Synonym(s):
LpDNPH S10 Cartridge Tube
About This Item
Recommended Products
Product Name
LpDNPH S10 Cartridge, bed wt. 350 mg, volume 3 mL, pkg of 50 ea (1 per nylon zip bag)
Agency
ASTM® D5197
EPA 100,IP-6A,TO-11A,TO-11A
NIOSH 2016
composition
bed wt., 350 mg
2 4-DNPH (High-purity silica gel)
packaging
pkg of 50 ea (1 per nylon zip bag)
technique(s)
active air sampling: suitable
volume
3 mL
application(s)
air monitoring
environmental
industrial hygiene
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service