Skip to Content
Merck
All Photos(8)

Documents

ZMS1199

Sigma-Aldrich

Anti-PSGL-1/CD162 Antibody, clone KPL-1 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

P-selectin glycoprotein ligand 1, PSGL-1 Selectin P ligand

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

KPL-1, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone KPL-1 is a ZooMAb® mouse monoclonal antibody that detects PSGL-1/CD162. It targets an epitope within the extracellular domain.

Immunogen

PSGL-1 transfected murine 300.19 pre B-cells.

Application

Quality Control Testing

Evaluated by Western Blotting in HL-60 cell lysates.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected PSGL-1/CD162 in HL-60 cell lysates.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PSGL-1/CD162 in lysate from Human peripheral blood mononuclear cells (PBMC)..

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected PSGL-1/CD162 in Human tonsil tissue sections.

Flow Cytometry Analysis: 0.1 µg from a representative lot detected PSGL-1/CD162 in one million Human peripheral blood mononuclear cells (PBMC).

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PSGL-1/CD162 in Jurkat cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

P-selectin glycoprotein ligand 1 (UniProt: Q14242; also known as PSGL-1 Selectin P ligand, CD162) is encoded by the SELPLG gene (Gene ID: 6404) in human. Leukocytes Express L-selectin ligands that are critical for leukocyte-leukocyte interactions at sites of inflammation. PSGL-1 is a disulfide-linked, homodimeric protein. It serves as a predominant leukocyte L-selectin ligand that displays appropriate sialyl Lewis x (sLex)-like carbohydrate determinants for L-selectin recognition. Its expression is observed in neutrophils, monocytes, and most lymphocytes. It is constitutively expressed on all human peripheral-blood T cells and is also expressed on skin-homing T cells. It is a single-pass, mucin-like type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-17), which is subsequently cleaved off to produce a form that contains an extracellular domain (18-320), a transmembrane domain (321-341), and a cytoplasmic domain (342-412). The mature form is generated following the cleaved of the propeptide (aa 18-41) region. PSGL-1 is a SLe(x)-type proteoglycan, which through high Affinity, calcium-dependent interactions with E-, P- and L-selectins, mediates rapid rolling of leukocytes over vascular surfaces during the initial steps in inflammation. Two isoforms of PSGL-1/CD162 have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals.  (Ref.: Tu, L., et al. (1999). J. Immunol. 163(9); 5070-5078; Fuhlbrigge, RC., et al. (1997). Nature. 389(6654); 978-981; Duivestijn, AM., et al. (1988). Am. J. Pathol. 130(1); 147-155).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
Express is a trademark of Merck KGaA, Darmstadt, Germany
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service