Key Documents
GBL460003
Grace Bio-Labs press fit tubing connectors
pack of 20 ea
Synonym(s):
press fit fitting, tubing connectors
Select a Size
20 EA
$50.40
$50.40
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
20 EA
$50.40
About This Item
UNSPSC Code:
41105108
NACRES:
NB.51
$50.40
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pack of 20 ea
manufacturer/tradename
Grace Bio-Labs 460003
General description
Press fit tubing connectors provide affordable and easy to use means to connect Grace′s chamber products to pumps and other devices for microfluidic applications.
Simply apply connectors by pressing the adhesive coated surface to the filling ports on the chamber surface and insert the appropriate length of tubing through the opening on the top. The simple hourglass design through the wall of the connector tightly secures the tubing to form a leak-proof seal in communication with the chamber. The open end of the tubing may be connected to a syringe or pump using a 25 gauge cannula or needle.
Choose from one of the most recent versions:
Certificates of Analysis (COA)