Key Documents
Z756903
Mouse restrainer with dorsal access
For mice 18-35 grams, pack of 1 ea
Synonym(s):
Rodent restrainer
Select a Size
1 EA
$96.10
$96.10
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onMay 06, 2025
Select a Size
Change View
1 EA
$96.10
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
$96.10
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onMay 06, 2025
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F46401-0000
General description
The accurate, safe and fast way to give injections. The dorsal skin of the mouse is easily accessed through the top circular opening so you can accurately perform subcutaneous injections without risk of being bitten and less likelihood of sticking your own finger.
  • Easy entry: draw mouse tail first or lead head first through the wide entry funnel into the back of the tube and then position stop gate in choice of two slots.
  • With tail first entry, tail is exposed at rear of tube allowing easy access for tattooing and tail bleeds
  • Overall Dimensions (L x W x H): 17.4 x 10 x 6.7cm (6.8 x 3.9 x 2.6 in.)
Choose from one of the most recent versions:
Certificates of Analysis (COA)