Key Documents
Z154563
Sigma-Aldrich
Positioning collar
PTFE/Glass composite, fits shaft o.d., 10 mm
Select a Size
1 EA
$86.00
$86.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$86.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
$86.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
PTFE gasket
feature
fits shaft o.d. 10 mm
manufacturer/tradename
Ace Glass 812710
General description
Designed for use with stirring shafts to position shaft in bearing and prevent it from dropping into flask. Gasket prevents scratching bearing and acts as a dust cover.
Choose from one of the most recent versions:
Certificates of Analysis (COA)