Skip to Content
Merck
All Photos(1)

Key Documents

Z688835

Nunc® UpCell Surface cell culture dish

Synonym(s):

cell culture dish, tissue culture dish

Sign Into View Organizational & Contract Pricing

Select a Size

30 EA
$1,670.00

$1,670.00


Estimated to ship onApril 11, 2025



Select a Size

Change View
30 EA
$1,670.00

About This Item

UNSPSC Code:
41121812
NACRES:
NB.22

$1,670.00


Estimated to ship onApril 11, 2025


material

polystyrene

sterility

sterile

feature

lid vented
membrane not included

packaging

case of 30 ea

manufacturer/tradename

Nunc 174903

dish diam. × H

60 mm × 15 mm

external L × W

60 mm × 15 mm

size

60 mm

surface area

21.5 cm2 , culture area (cm2)/well

working volume

5 mL

binding type

(UpCell Surface)

Looking for similar products? Visit Product Comparison Guide

General description

The Nunc UpCell Surface dishes have a temperature responsive cell culture surface. These UpCell Surface dishes are suitable for harvesting of cells with intact surface proteins, for culture passaging, single-cell analyses and cell transplantation research. The UpCell surface enables harvesting of cell sheets and creation of 3D tissue models which are held together by normal cell junctions and extracellular matrix.


  • Harvesting of cells with intact surface proteins
  • No trypsinization - preserve cell surface proteins
  • No physical force - get high cell viability
  • Releases adherent cells by reduction of temperature of the cell culture
  • For culture passaging, single-cell analyses and cell transplantation research
  • Enables harvesting of cell sheets and creations of 3D tissue models held together by normal cell junctions and extracellular matrix
  • Minimal hands-on time
  • For research and single-use only
  • It is quick, clean and simple-just reduce the temperature

Note: Suitable for the harvesting of cells with intact surface proteins, for cell culture passage, single-cell analyses and cell transplantation research

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
UpCell is a trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. How many minutes does it take to Detach cells at 20 degree from 100 mm dish?  Does cells be released as a single cells ? Or sheet format?  Your answer is appreciated. Thanks 

    1 answer
    1. This is a product Nunc/ThermoFisher. The manufacturer offers the same detachment information, regardless of dish size or number of wells.
      The following example protocol for cell suspensions:
      1. Mouse Peritoneal Macrophages were washed with Ca2+– and Mg2+–free PBS to remove non-adherant cells
      2. Fresh medium including FBS was added to the plate.
      3. Cultures were incubated for 30 minutes at 20°C prior to harvesting.

      The following example protocol for harvesting cells as a single sheet:
      Madin-Darby Canine Kidney (MDCK) cells in Dulbecco’s Modifi ed Eagle Medium (DMEM) supplemented with 10% fetal bovine serum (FBS), penicillin (100 units/mL) and streptomycin (100 μg/mL) were seeded at 5.7 x 104 cell/cm2 in a 3.5 cm dish with UpCell Surface. The cells were incubated for 21 days at 37°C in a humidifi ed atmosphere of 5% CO2 in air. The cells were then harvested as an intact sheet:
      1. The dish was placed at 20°C for 60 min.
      2. A supporting membrane was placed on top of the cell layer and the culture medium was aspirated
      3. The edge of the membrane was gently released from the dish using forceps, and the membrane with the attached cell layer was then transferred to a cell culture insert and incubated at 20°C for 30 min.
      4. Sufficient culture medium was added to cover the cell sheet and membrane and the membrane was removed from the cell layer using forceps

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service