Key Documents
Z117242
Sigma-Aldrich
Aldrich® dry ice condenser for Büchi® rotary evaporators
with hose barb
Select a Size
1 EA
$893.00
$893.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$893.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$893.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
male joint SJ35/20
volume measuring range
500 mL (dry ice reservoir)
Related Categories
General description
For Büchi rotary evaporators R110, and R151. For use with short stem duct.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)