Z672521
BRAND® seripettor® pro bottle-top dispenser
volume 2.5 - 25 mL
Synonym(s):
Bottle-Top Dispenser
Select a Size
$1,460.00
Estimated to ship onMay 21, 2025
Select a Size
About This Item
$1,460.00
Estimated to ship onMay 21, 2025
Recommended Products
feature
subdivision 0.5 mL
manufacturer/tradename
BRAND 4720450
volume
2.5 - 25 mL
fitting
(GL 45/32)
(GL 45/38)
(GL 45/S 40)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Not compatible with hydrofluoric acid, or other chemicals that attack polypropylene, PTFE, ETFE, FEP, PFA, alumina-ceramic, platinum-iridium, borosilicate glass and carbon disulfide
Features and Benefits
- Low-cost replaceable dispensing cartridge with wiping seal - one spare dispensing cartridge included
- Semi-automatic dispensing - The spring loaded piston refills the cylinder after dispensing
- Volume setting - Scalloped track allows for quick and exact setting of desired volume
- Disassembles without tools - All parts can be replaced quickly without tools
- The innovate design makes cleaning and maintenance easy
- Supplied with discharge tube, filling tube, spare dispensing cartridge and adapters, PP (GL 45/32 and GL 45/540)
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service