Key Documents
BR4600361
BRAND® Dispensette® S Digital bottle-top dispenser
volume range 5-50 mL, with recirculation valve
Synonym(s):
bottle top dispensers
Select a Size
1 EA
$1,160.00
$1,160.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$1,160.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
$1,160.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
feature
incremental 0.2 mL
DE-M marking
with recirculation valve
manufacturer/tradename
BRAND 4600361
volume range
5-50 mL
fitting
(GL 32-33)
GL 38
S 40
General description
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
Features and Benefits
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Other Notes
Digital-The mechanical volume setting is easy to read and ensures accurate and reproducible volume control. Easy Calibration Technique for calibration adjustment in seconds without tools. A calibration flag indicates changes from factory settings.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Related Content
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.