Key Documents
BR4600140
BRAND® Dispensette® S Analog-adjustable bottle-top dispenser
volume range 1-10 mL, without recirculation valve
Synonym(s):
bottle top dispensers
Select a Size
1 EA
$714.00
$714.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$714.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
$714.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
feature
incremental 0.2 mL
DE-M marking
without recirculation valve
manufacturer/tradename
BRAND 4600140
volume range
1-10 mL
fitting
(GL 28/S28)
GL 32
GL 38
S 40
General description
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
Features and Benefits
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Other Notes
Analog-adjustable - Rapid and safe volume setting with new internal scalloped track. Simple calibration adjustment with a supplied tool.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Related Content
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.