Skip to Content
Merck
All Photos(1)

Key Documents

Z358762

Culture dishes

size 100 mm × 25 mm, sterile, polystyrene

Synonym(s):

cell culture dish, microbiology dish, petri dish

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
$66.50
500 EA
$1,230.00

$66.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
20 EA
$66.50
500 EA
$1,230.00

About This Item

UNSPSC Code:
41121812
NACRES:
NB.22

$66.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

polystyrene
round

sterility

sterile

feature

lid

packaging

pack of 20 ea
pkg of 500 ea

dish capacity

125 mL

size

100 mm × 25 mm

surface area

55 cm

application(s)

agriculture

binding type

non-treated surface

General description

Clear, with stacking features
Designed with extra depth for plant tissue culture

Application

Culture dishes are suitable for routine procedures and automated processes.

Features and Benefits

Culture dishes provide the following benefits:
  • Made of biomedical polystyrene for clear visibilty of cultures and avoids distortion.
  • Optically clear and precision-molded.
  • Prevents condensation due to the presence of venting ribs.
  • Non-vented lids for stable stacking.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David C Butler et al.
PloS one, 6(12), e29199-e29199 (2012-01-05)
Huntington's disease (HD) is a fatal autosomal dominant neurodegenerative disorder caused by a trinucleotide (CAG)(n) repeat expansion in the coding sequence of the huntingtin gene, and an expanded polyglutamine (>37Q) tract in the protein. This results in misfolding and accumulation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service