Skip to Content
Merck
All Photos(1)

Key Documents

C0542

Magenta vessel

GA-7/GA-7-3 cover

Synonym(s):

Laboratory Vessel, Research Vessel

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
$83.20
100 EA
$248.00

$83.20


Estimated to ship onMay 29, 2025



Select a Size

Change View
25 EA
$83.20
100 EA
$248.00

About This Item

UNSPSC Code:
41161500
NACRES:
NB.22

$83.20


Estimated to ship onMay 29, 2025


material

polypropylene

description

Autoclavable

W × L

77 mm (3 in.) × 77 mm (3 in.)

application(s)

agriculture

compatibility

closure fits either V8380 or V8505

Looking for similar products? Visit Product Comparison Guide

Application

Magenta vessel has been used in microcosm construction and assembling. It is suitable for plant tissue culture, multiplication, transport, and pre-transplanting.

Features and Benefits

  • Promotes quick seed germination and easier inoculation and transfer.
  • High relative growing area and low incubation shelving cost.
  • Designed for improved stacking and to save space during autoclaving, shipping & storage.
  • Excellent plant protection - no breakage or leakage.
  • Autoclavable and reusable, resulting in a low cost per plant.
  • Reduces contamination and the resulting profit loss.
  • Convenient to operate and minimum handling when used with MAGENTA 7-Way Tray.
  • Lightweight.

Legal Information

Magenta is a trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

1–2 of 2 Questions  
  1. Good morning, Which is the capacity of the magenta vessel CO542?

    1 answer
    1. This product is only the lid for the Magenta GA vessels. The GA7-3 vessel, product V8380, has a calculated maximum capacity of of 442 mL. The GA-7 vessel, product S8505, has a calculated maximum volume of 589 mL. However, these vessels are typically used for plant cell and tissue culture. The working volume may vary based on the plant species and tissue type. Most applications will require sufficient head space for growth; generally 25 - 50% of the available volume.

      Helpful?

  2. the CO542 Magenta Vessel does that include the cover with the tray?

    1 answer
    1. This product includes the cover only, without the vessel or tray. This cover will fit both the GA-7 and the GA-3 vessels.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service