Skip to Content
Merck
All Photos(1)

Key Documents

Z331910

BRAND® Dispensette® III bottle-top dispenser, digital, Easy calibration

5-50 mL, with SafetyPrime

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500

feature

with SafetyPrime

manufacturer/tradename

BRAND 4700361 EU
BrandTech 4701361 US

measuring range

5-50 mL

Looking for similar products? Visit Product Comparison Guide

General description

Dispensette III (red color-code): for dispensing aggressive reagents, including concentrated acids such as H3PO4, H2SO4, bases like NaOH, KOH, saline solutions as well as many organic solvents - directly from a supply bottle.

  • Easy to dismantle for cleaning, with replaceable filling valves
  • Autoclavable at 121°C
  • Digital display: Especially easy to read and dispensing volume can be set accurately and reproducibly (mechanical counter)
  • Easy Calibration: Innovative technique for adjustment in seconds without tools
  • SafetyPrime recirculation valve reduces reagent waste and the safety discharge system reduces the risk of inadvertent dispensing and splashes
  • To allow the bottle label to face the user, the valve block can be rotated 360°
  • A telescopic filling tube adjusts easily to different size bottles

The parts of the Dispensette Organic which are in contact with medium are made of: Borosilicate glass, ceramic, tantalum, ETFE, FEP & PFA. The discharge tube safety screw cap is PP.

Operating limits:
  • Vapor pressure max. 500 mbar
  • Viscosity max. 500 mm2/s
  • Temperature max. 40°C
  • Density max. 2.2 g/cm3
  • Accuracy: ±0.5%, precision ±0.1%

Linkage

See the Compatibility Chart to choose the correct bottle-top dispenser.

Other Notes

BR4700361 (India)

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
SafetyPrime is a trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service