Z331740
BRAND® pipette
withdraw volume 3 mL (including bulb), graduations: no, drop quantity 25 per mL
Synonym(s):
LAB Pipette
Select a Size
$85.60
Select a Size
About This Item
$85.60
Recommended Products
material
PE-LD
sterility
non-sterile
feature
graduations: no
packaging
pack of 500
pkg of 500 ea
manufacturer/tradename
BRAND 747750
withdraw volume measuring range
3 mL (including bulb)
L
152 mm
tip O.D.
2.8 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Please note: can be sterilized by gas or γ-irradiation
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service