Key Documents
Z685127
Ace powder funnel with flat side
top diam. 100 mm, joint: ST/NS 24/40
Select a Size
1 EA
$215.00
$215.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$215.00
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
$215.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass
Joint
joint: ST/NS24/40
manufacturer/tradename
Ace Glass
top diam.
100 mm
Related Categories
General description
Ace powder funnel with standard taper joint and one flattened side for easy of use with multi-neck flasks.
Choose from one of the most recent versions:
Certificates of Analysis (COA)