Skip to Content
Merck
All Photos(1)

Key Documents

CLS7095D5X

Corning® disposable Pasteur pipettes, bulk pack, non-sterile, unplugged

size 5.75 in., borosilicate

Synonym(s):

pipettes, serological pipets, serological pipettes

Sign Into View Organizational & Contract Pricing

Select a Size

200 EA
$80.50
1000 EA
$93.60

$80.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
200 EA
$80.50
1000 EA
$93.60

About This Item

UNSPSC Code:
41121500
eCl@ss:
32079210
NACRES:
NB.04

$80.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

borosilicate glass

sterility

non-sterile

packaging

case of 1000 ea
pack of 200 ea

manufacturer/tradename

Corning 7095D-5X

size

5.75 in.

application(s)

PFAS testing

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® disposable Pasteur pipettes are made of sturdy and optically clear borosilicate glass.
  • Single-use pipettes prevent the risk of cross-contamination of valuable specimens or laboratory reagents.
  • Feature a constriction one inch below the top to facilitate plugging.
  • Ideal for use in non-volumetric transfer work in bacteriology, immunology, hematology, and serology studies, as well as blood bank and clinical chemistry procedures.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service